Learn More
CD95 Mouse anti-Human, PE, Clone: DX2, BD
Mouse Monoclonal Antibody
451.00€ - 479.00€
Spécification
Antigène | CD95 |
---|---|
Clone | DX2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | PE |
Produktcode | Marke | Quantité | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantité | Preis | Menge & Verfügbarkeit | |||||
15838768
|
BD Biosciences
556641 |
50 Tests |
451.00€
1 pièce |
Expédition estimée: 17-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15878348
|
BD Biosciences
555674 |
100 Tests |
479.00€
1 pièce |
Expédition estimée: 17-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Beschreibung
Reacts with the human form of the 40-50kDa transmembrane cell surface Fas or Apo-1 antigen. The Fas/Apo-1 antigen is a polypeptide which plays a role in the programmed sequence of events leading to cell death, termed apoptosis. DX2 cross-reacts with a subset of peripheral blood lymphocytes, monocytes and granulocytes of baboon and both rhesus and cynomolgus macaque monkeys. The distribution on lymphocytes is similar to that seen with human peripheral blood lymphocytes being broadly expressed on all major lymphocyte subsets. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: DX2
Isotype: IgG1, κ
Cross Reactivity: Baboon, Cynomolgus
Flow Cytometry
Spezifikation
CD95 | |
Flow Cytometry | |
PE | |
RUO | |
Baboon, Canine, Cynomolgus, Human, Porcine, Rhesus Monkey | |
Human CD95-transfected L Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
DX2 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
APO-1; FAS; TNFRSF6; APT1; ALPS1A; FAS1; FASTM; FASLG receptor |
For Research Use Only.