missing translation for 'onlineSavingsMsg'
Learn More

CDK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10122-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16806303

  • 112.71€ / 20µL

Expédition estimée
26-02-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as does cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.
Spécification

Spécification

CDK2
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P24941, P97377, Q63699
CDK2, Cell division protein kinase 2, cyclin dependent kinase 2, p33 protein kinase, p33(CDK2)
Rabbit
IgG
20 μL
-20°C
Primary
1017, 12566, 362817
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Flow Cytometry
Unconjugated
CDK2
Liquid
Cdk2
CDK2 Fusion Protein Ag0166
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat, Mouse
Vidéos
FDS
Documents