missing translation for 'onlineSavingsMsg'
Learn More

CDK3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 55103-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16806555

  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The antibody is specific to CDK3.

This gene encodes a member of the cyclin-dependent protein kinase family. The protein promotes entry into S phase, in part by activating members of the E2F family of transcription factors. The protein also associates with cyclin C and phosphorylates the retinoblastoma 1 protein to promote exit from G0.
Spécification

Spécification

CDK3
Polyclonal
Unconjugated
CDK3
CDK3, Cell division protein kinase 3, cyclin dependent kinase 3
Rabbit
Antigen Affinity Chromatography
RUO
1018, 69681
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.8 mg/mL
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
Q00526, Q80YP0
Cdk3
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions