missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDKAL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22988-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody recognizes the 65 kDa isoform of CDKAL1.
The protein encoded by this gene is a member of the methylthiotransferase family. The function of this gene is not known. Genome-wide association studies have linked single nucleotide polymorphisms in an intron of this gene with susceptibility to type 2 diabetes.Spécification
CDKAL1 | |
Polyclonal | |
Unconjugated | |
CDKAL1 | |
CDKAL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54901, 68916 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5VV42, Q91WE6 | |
CDKAL1 | |
CDKAL1 Fusion Protein Ag19060 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |