missing translation for 'onlineSavingsMsg'
Learn More

CEL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15384-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16804544

  • 329.85€ / 150µL

Expédition estimée
02-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CEL catalyzes fat and vitamin absorption. CEL acts in concert with pancreatic lipase and colipase for the complete digestion of dietary triglycerides.The protein encoded by this gene is a glycoprotein secreted from the pancreas into the digestive tract and from the lactating mammary gland into human milk. The physiological role of this protein is in cholesterol and lipid-soluble vitamin ester hydrolysis and absorption. This encoded protein promotes large chylomicron production in the intestine. Also its presence in plasma suggests its interactions with cholesterol and oxidized lipoproteins to modulate the progression of atherosclerosis. In pancreatic tumoral cells, this encoded protein is thought to be sequestrated within the Golgi compartment and is probably not secreted. This gene contains a variable number of tandem repeat (VNTR) polymorphism in the coding region that may influence the function of the encoded protein.
Spécification

Spécification

CEL
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07882, P19835, Q64285
BAL, Bile salt stimulated lipase, Bile salt-activated lipase, BSDL, BSSL, Bucelipase, Carboxyl ester lipase, CEase, CEL, CELL, Cholesterol esterase, FAP, FAPP, LIPA, MODY8, Pancreatic lysophospholipase, Sterol esterase
Rabbit
IgG
150 μL
-20°C
Primary
1056, 12613, 24254
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CEL
Liquid
CEL
CEL Fusion Protein Ag5262
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents