Learn More
Eppendorf Cell Imaging Slides
Convenient, reliable, chemically resistant slides suitable for fixation protocols.
Marque:  Eppendorf 0030742044
Code nomenclature Nacres: NB.13
Informations supplémentaires : Poids : 297.00000kg
Code produit. 15660547
  • 130.50€ / 16 pièces
Expédition estimée
12-07-2019
Ajouter au panier
Description
Description
  • 1 mm-thick slide with excellent signal-to-noise ratios for fluorescent staining protocols
  • Facilitate growth of most adherent cell types due to innovative TC-treated glass surface
  • High chemical resistance, so fixation even with acetone is possible without problems
  • Benefit from easy and tool-free removal of the chambers with minimized glue residues
  • Precise planarity supports high resolution microscopes and autofocus systems
Spécification
Spécification
1
Non-cytotoxic, sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free.
Yes
Individually wrapped
Tissue-Culture Treated
Yes
16
Sterile
Vidéos