missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CES1 (Human) Recombinant Protein
Human CES1 full-length ORF ( AAH12418, 1 a.a. - 566 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00001066-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: MWLPALVLATLAASAAWGHPSSPPVVDTVHGKVLGKFVSLEGFAQPVAIFLGIPFAKPPLGPLRFTPPQPAEPWSFVKNATSYPPMCTQDPKAGQLLSELFTNRKENIPLKLSEDCLYLNIYTPADLTKKNRLPVMVWIHGGGLMVGAASTYDGLALAAHENVVVVTIQYRLGTWGFFSTGDEHSRGNWGHLDQVAALRWVQDNIASFGGNPGSVTIFGESAGGESVSVLVLSPLAKNLFHRAISESGVALTSVLVKKGDVKPLAEQIAITAGCKTTTSAVMVHCLRQKTEEELLETTLKMKFLSLDLQGDPRESQPLLGTVIDGMLLLKTPEELQAERNFHTVPYMVGINKQEFGWLIPMLMSYPLSEGQLDQKTAMSLLWKSYPLVCIAKELIPEATEKYLGGTDDTVKKKDLFLDLIADVMFGVPSVIVARNHRDAGAPTYMYEFQYRPSFSSDMKPKTVIGDHGDELFSVFGAPFLKEGASEEEIRLSKMVMKFWANFARNGNPNGEGLPHWPEYNQKEGYLQIGANTQAAQKLKDKEVAFWTNLFAKKAVEKPPQTEHIEL
Spécification
AAH12418 | |
carboxylesterase 1 (monocyte/macrophage serine esterase 1) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CES1 | |
GST |
1066 | |
Wheat germ expression system | |
25 μg | |
ACAT, CEH, CES2, HMSE, HMSE1, MGC117365, PCE-1, SES1, TGH | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |