missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CETP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13459-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Cholesteryl ester transfer protein (CETP) transfers cholesteryl esters between lipoproteins. CETP may effect susceptibility to atherosclerosis.Spécification
CETP | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11597 | |
CETP, HDLCQ10, Lipid transfer protein I | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
1071 |
Western Blot | |
Unconjugated | |
CETP | |
Liquid | |
CETP | |
CETP Fusion Protein Ag3923 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |