missing translation for 'onlineSavingsMsg'
Learn More

Chloropentafluorobenzène, +98 %, Thermo Scientific Chemicals

Catalog Number p-7022550
Click to view available options
Quantité:
10 g
50 g
250 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
344-07-0
C6ClF5
202.51
MFCD00000534
KGCDGLXSBHJAHZ-UHFFFAOYSA-N
chloropentafluorobenzene, pentafluorochlorobenzene, benzene, chloropentafluoro, pentafluorophenyl chloride, chloroperfluorobenzene, ccris 2394, pentafluorochloro benzene, 1-chloro-2,3,4,5,6-pentafluoro-benzene, benzene, 1-chloro-2,3,4,5,6-pentafluoro, pentafluorophenylchloride
9579
1-Chloro-2,3,4,5,6-pentafluorobenzène
FC1=C(F)C(F)=C(Cl)C(F)=C1F
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
344-07-0
C6ClF5
202.51
MFCD00000534
KGCDGLXSBHJAHZ-UHFFFAOYSA-N
chloropentafluorobenzene, pentafluorochlorobenzene, benzene, chloropentafluoro, pentafluorophenyl chloride, chloroperfluorobenzene, ccris 2394, pentafluorochloro benzene, 1-chloro-2,3,4,5,6-pentafluoro-benzene, benzene, 1-chloro-2,3,4,5,6-pentafluoro, pentafluorophenylchloride
9579
1-Chloro-2,3,4,5,6-pentafluorobenzène
FC1=C(F)C(F)=C(Cl)C(F)=C1F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Chloropentafluorobenzene
Densité 1.568
Point d’ébullition 116°C to 118°C
Indice de réfraction 1.424
Quantité 10 g
Beilstein 1819389
Poids de la formule 202.51
Pourcentage de pureté ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'