Learn More
CHRFAM7A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21586-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody, raised against 241-386 amino acids of human CHRFAM7A protein, recognizes both CHRNA7 and CHRFAM7A.
The nicotinic acetylcholine receptors (nAChRs) are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The family member CHRNA7, which is located on chromosome 15 in a region associated with several neuropsychiatric disorders, is partially duplicated and forms a hybrid with a novel gene from the family with sequence similarity 7 (FAM7A). Alternative splicing has been observed, and two variants exist, for this hybrid gene. The N-terminally truncated products predicted by the largest open reading frames for each variant would lack the majority of the neurotransmitter-gated ion-channel ligand binding domain but retain the transmembrane region that forms the ion channel. Although current evidence supports transcription of this hybrid gene, translation of the nicotinic acetylcholine receptor-like protein-encoding open reading frames has not been confirmed.Spécification
CHRFAM7A | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q494W8 | |
CHRFAM7A, CHRNA7, CHRNA7 DR1, CHRNA7 FAM7A fusion protein, D 10 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
89832 |
Western Blot | |
Unconjugated | |
CHRFAM7A | |
Liquid | |
CHRFAM7A | |
CHRFAM7A Fusion Protein Ag16151 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |