missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIP29 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15798-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein that is upregulated in response to various cytokines. The encoded protein may play a role in cell cycle progression. A translocation between this gene and the myeloid/lymphoid leukemia gene, resulting in expression of a chimeric protein, has been associated with acute myelomonocytic leukemia. Pseudogenes exist on chromosomes 7 and 8. Alternatively spliced transcript variants have been described.Spécification
CIP29 | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P82979, Q498U4, Q9D1J3 | |
CIP29, HCC1, HSPC316, Nuclear protein Hcc 1, SARNP | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
362819, 66118, 84324 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
SARNP | |
Liquid | |
Sarnp | |
CIP29 Fusion Protein Ag8507 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |