missing translation for 'onlineSavingsMsg'
Learn More

cis-4-(boc-amino)cyclohexaneacétaldéhyde, 97 %, Thermo Scientific Chemicals

Catalog Number p-7041435
Click to view available options
Quantité:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
847417-37-2
C13H23NO3
241.331
MFCD03844591
OPGBSEXLSWYFOR-UHFFFAOYSA-N
tert-butyl trans-4-2-oxoethyl cyclohexylcarbamate, tert-butyl cis-4-2-oxoethyl cyclohexylcarbamate, tert-butyl n-4-2-oxoethyl cyclohexyl carbamate, carbamic acid,n-trans-4-2-oxoethyl cyclohexyl-, 1,1-dimethylethyl ester, opgbsexlswyfor-xypyzodxsa-n, tert-butyl 4-2-oxoethyl cyclohexyl carbamate, cis-tert-butyl 4-2-oxoethyl cyclohexyl carbamate, trans-tert-butyl-4-2-oxoethyl-cyclohexylcarbamate, cis-4-tert-butoxycarbonylamino cyclohexylacetaldehyde, tert-butyl trans-4-2-oxoethyl cyclohexyl carbamate
24720884
Tert-butyl N-[4-(2-oxoéthyl)cyclohexyl]carbamate
CC(C)(C)OC(=O)NC1CCC(CC1)CC=O
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
847417-37-2
C13H23NO3
241.331
MFCD03844591
OPGBSEXLSWYFOR-UHFFFAOYSA-N
tert-butyl trans-4-2-oxoethyl cyclohexylcarbamate, tert-butyl cis-4-2-oxoethyl cyclohexylcarbamate, tert-butyl n-4-2-oxoethyl cyclohexyl carbamate, carbamic acid,n-trans-4-2-oxoethyl cyclohexyl-, 1,1-dimethylethyl ester, opgbsexlswyfor-xypyzodxsa-n, tert-butyl 4-2-oxoethyl cyclohexyl carbamate, cis-tert-butyl 4-2-oxoethyl cyclohexyl carbamate, trans-tert-butyl-4-2-oxoethyl-cyclohexylcarbamate, cis-4-tert-butoxycarbonylamino cyclohexylacetaldehyde, tert-butyl trans-4-2-oxoethyl cyclohexyl carbamate
24720884
Tert-butyl N-[4-(2-oxoéthyl)cyclohexyl]carbamate
CC(C)(C)OC(=O)NC1CCC(CC1)CC=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau cis-4-(Boc-amino)cyclohexaneacetaldehyde
Poids de la formule 241.32
Pourcentage de pureté 97%
Quantité 250 mg

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'