missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNIH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13144-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Regulates the trafficking and gating properties of AMPA- selective glutamate receptors (AMPARs). Promotes their targeting to the cell membrane and synapses and modulates their gating properties by regulating their rates of activation, deactivation and desensitization.Spécification
CNIH3 | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D0Q0Y7, Q6ZWS4, Q8TBE1 | |
CNIH3, Protein cornichon homolog 3 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
149111, 690252, 72978 |
Western Blot | |
Unconjugated | |
CNIH3 | |
Liquid | |
Cnih3 | |
CNIH3 Fusion Protein Ag3648 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |