missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13427-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.Spécification
CNPase | |
Polyclonal | |
Unconjugated | |
CNP | |
CNP, CNP1, CNPase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1267, 12799, 25275 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09543, P13233, P16330 | |
CNP | |
CNPase Fusion Protein Ag4227 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |