missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTNAP3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27410-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The specific function of this protein remains unknown. The protein encoded by this gene belongs to the NCP family of cell-recognition molecules. This family represents a distinct subgroup of the neurexins. NCP proteins mediate neuron-glial interactions in vertebrates and glial-glial contact in invertebrates. The protein encoded by this gene may play a role in cell recognition within the nervous system. Alternatively spliced transcript variants encoding different isoforms have been described but their biological nature has not been determined.Spécification
CNTNAP3 | |
Polyclonal | |
Unconjugated | |
CNTNAP3 | |
CASPR3, CNTNAP3, CNTNAP3A, KIAA1714, RP11 138L21.1, RP11 290L7.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79937 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BZ76 | |
CNTNAP3 | |
CNTNAP3 Fusion Protein Ag26624 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |