missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16654-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Component of the coenzyme Q biosynthetic pathway. May play a role in organizing a multi-subunit COQ enzyme complex required for coenzyme Q biosynthesis. Required for steady-state levels of other COQ polypeptides.Spécification
COQ4 | |
Polyclonal | |
Unconjugated | |
COQ4 | |
CGI 92, COQ4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227683, 366013, 51117 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4FZU1, Q8BGB8, Q9Y3A0 | |
COQ4 | |
COQ4 Fusion Protein Ag10089 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |