missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPEB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13274-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody may recognize several bands, but only ∽56 kDa band is the right one, other (40∽50 kDa) is un-specific bands.
This gene encodes a protein which, with 5-lipoxygenase, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the nuclear membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the nuclear membrane and inhibit 5-lipoxygenase activation. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Spécification
CPEB1 | |
Polyclonal | |
Unconjugated | |
CPEB1 | |
CEBP, CPE binding protein 1, CPE BP1, CPEB, CPEB1, h CEBP, hCPEB 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12877, 293056, 64506 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P0C279, P70166, Q9BZB8 | |
CPEB1 | |
CPEB1 Fusion Protein Ag3994 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |