missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13385-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
CPN1 is a plasma metallo-protease that cleaves basic amino acids from the C terminal of peptides and proteins. The protein is important in the regulation of peptides like kinins and anaphylatoxins, and has also been known as kininase-1 and anaphylatoxin inactivator. This protein is a tetramer comprised of two identical regulatory subunits and two identical catalytic subunits.Spécification
CPN1 | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15169, Q9EQV8, Q9JJN5 | |
ACBP, Anaphylatoxin inactivator, Arginine carboxypeptidase, CPN, CPN1, Kininase 1, Lysine carboxypeptidase, Plasma carboxypeptidase B, SCPN, Serum carboxypeptidase N | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
1369, 365466, 93721 |
Immunocytochemistry, Immunofluorescence, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
CPN1 | |
Liquid | |
Cpn1 | |
CPN1 Fusion Protein Ag4035 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Rat, Human |