Learn More
CPZ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène CPZ
Concentration 0.22 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications
Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16815354
Afficher les documents Détails de la promotion
Proteintech
15944-1-AP-20UL
20 μL
140.00€
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 03-05-2024
pour voir le stock disponible 
16805354
missing translation for 'viewDocuments' Promotion Details
Proteintech
15944-1-AP-150UL
150 μL
399.00€
150µL
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 03-05-2024
Description
CPZ is a member of the metallocarboxypeptidase family. This enzyme displays carboxypeptidase activity towards substrates with basic C-terminal residues. It is most active at neutral pH and is inhibited by active site-directed inhibitors of metallocarboxypeptidases.
Specifications
Specifications
CPZ
Western Blot
Unconjugated
Rabbit
Mouse, Human, Rat
O54858, Q66K79, Q8R4V4
242939, 83575, 8532
CPZ Fusion Protein Ag8429
Primary
-20°C
CPZ
0.22 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
carboxypeptidase Z, CPZ
CPZ
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents
Documents
Certificates