Learn More
Abeba™ Crawler ALU 34813 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Marque:  Abeba™ 34813/41
Code nomenclature Nacres: HA.03
Informations supplémentaires : Poids : 1.24700kg
Code produit. 15660078
  • 119.20€ / 1 paire
Expédition estimée
17-10-2019
Ajouter au panier
Description
Description
  • CE EN ISO 20345:2011 S1P SRC
  • Microfiber with breathable textile inlays
  • Black/Yellow
  • Aluminum toe cap
  • Foamed PU midsole
  • Metalfree penetration resistance
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Discontinued model
Spécification
Spécification
CE EN ISO 20345:2011 S1P SRC
Microfiber
Slip resistant
34813
Black, Yellow
41
Polyurethane
Aluminum