Learn More
Abeba™ Crawler ALU 4502 Shoes
Aluminum toe cap provides lightweight toe protection. Sanitized™ treated.
Marque:  Abeba™ 4502/39
Code nomenclature Nacres: HA.03
Informations supplémentaires : Poids : 1.24000kg
Code produit. 15661528
  • 109.00€ / 1 paire
Expédition estimée
18-09-2019
Ajouter au panier
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber black/red
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Upper one piece
Spécification
Spécification
CE EN ISO 20345:2011 S2 SRC
Microfiber
Slip resistant
4502
Black
39
Polyurethane
Aluminum