Learn More
Abeba™ Crawler ALU 4572 Shoes
Aluminum toe cap provides lightweight toe protection. Sanitized™ treated.
Marque:  Abeba™ 4572/45
Code nomenclature Nacres: HA.03
Informations supplémentaires : Poids : 1.24800kg
Code produit. 15661638
  • 114.40€ / 1 paire
Expédition estimée
16-07-2019
Ajouter au panier
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Velours with breathable textile inlays
  • Black/yellow
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • EXPIRED size 36 - 40
  • Scratch-resistant coating in toe and heel area
Spécification
Spécification
CE EN ISO 20345:2011 S1 SRC
Velours
Slip resistant
4572
Black
45
Polyurethane
Aluminum
Vidéos