missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRMP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14686-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, neuronal growth cone collapse and cell migration.Spécification
CRMP2 | |
Polyclonal | |
Unconjugated | |
DPYSL2 | |
CRMP 2, CRMP2, DHPRP2, dihydropyrimidinase like 2, DPYSL2, DRP 2, DRP2, N2A3, ULIP 2, ULIP2, Unc 33 like phosphoprotein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12934, 1808, 25416 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.64 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08553, P47942, Q16555 | |
DPYSL2 | |
CRMP2 Fusion Protein Ag5985 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |