missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CROT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13543-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody can bind the close sequences genes.
CROT encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4, 8-dimethylnooyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described.Spécification
CROT | |
Polyclonal | |
Unconjugated | |
CROT | |
COT, CROT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54677, 74114, 83842 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11466, Q9DC50, Q9UKG9 | |
CROT | |
CROT Fusion Protein Ag4486 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |