missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CROT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigène | CROT |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry |
Classification | Polyclonal |
Conjugué | Unconjugated |
Product Code | Brand | Quantité | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantité | Price | Quantity & Availability | |||||
16801814
|
Proteintech
13543-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 06-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16891804
|
Proteintech
13543-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 06-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This antibody can bind the close sequences genes.
CROT encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4, 8-dimethylnooyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described.Specifications
CROT | |
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P11466, Q9DC50, Q9UKG9 | |
54677, 74114, 83842 | |
CROT Fusion Protein Ag4486 | |
Primary | |
-20°C | |
CROT |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
COT, CROT | |
CROT | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |