missing translation for 'onlineSavingsMsg'
Learn More

CSAD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 22870-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802125

  • 112.71€ / 20µL

Expédition estimée
10-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the group 2 decarboxylase family. A similar protein in rodents plays a role in multiple biological processes as the rate-limiting enzyme in taurine biosynthesis, catalyzing the decarboxylation of cysteinesulfinate to hypotaurine. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Spécification

Spécification

CSAD
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q64611, Q9DBE0, Q9Y600
CSAD, CSD, PCAP, Sulfinoalanine decarboxylase
Rabbit
IgG
20 μL
-20°C
Primary
246277, 51380, 60356
Immunohistochemistry (Paraffin)
Unconjugated
CSAD
Liquid
CSAD
CSAD Fusion Protein Ag15828
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Vidéos
FDS
Documents