missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CWC25 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24416-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The function of this protein remains unknown.Spécification
CWC25 | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9DBF7, Q9NXE8 | |
CCDC49, CWC25 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
54883, 67480 |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
CWC25 | |
Liquid | |
CWC25 | |
CWC25 Fusion Protein Ag17037 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |