missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26756-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Chemokines play important roles in inflammation and critical for the recruitment of effector immune cells to sites of infection. Historically, CD183 was the third CXC chemokine receptor discovered and therefore, commonly designated as CXCR3. Binding of chemokines to CD183 induces cellular responses that are involved in leukocyte traffic, most notably integrin activation, cytoskeletal changes and chemotactic migration. The CXC chemokine receptor (CXCR) 3, a G-protein-coupled 7-transmembrane receptor, is expressed at high levels on activated and memory T cells, B cells, natural killer cells, and plasmacytoid monocytes.Spécification
CXCR3 | |
Polyclonal | |
Unconjugated | |
CXCR3 | |
CD183, CKR L2, CMKAR3, CXC R3, CXCR 3, CXCR3, CXCR3A, CXCR3B, G protein coupled receptor 9, GPR9, IP 10 receptor, IP10 R, Mig R, MigR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12766, 2833 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88410, P49682 | |
CXCR3 | |
CXCR3 Fusion Protein Ag25056 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |