missing translation for 'onlineSavingsMsg'
Learn More

CXCR3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26756-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16835125

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Chemokines play important roles in inflammation and critical for the recruitment of effector immune cells to sites of infection. Historically, CD183 was the third CXC chemokine receptor discovered and therefore, commonly designated as CXCR3. Binding of chemokines to CD183 induces cellular responses that are involved in leukocyte traffic, most notably integrin activation, cytoskeletal changes and chemotactic migration. The CXC chemokine receptor (CXCR) 3, a G-protein-coupled 7-transmembrane receptor, is expressed at high levels on activated and memory T cells, B cells, natural killer cells, and plasmacytoid monocytes.
Spécification

Spécification

CXCR3
Polyclonal
Unconjugated
CXCR3
CD183, CKR L2, CMKAR3, CXC R3, CXCR 3, CXCR3, CXCR3A, CXCR3B, G protein coupled receptor 9, GPR9, IP 10 receptor, IP10 R, Mig R, MigR
Rabbit
Antigen Affinity Chromatography
RUO
12766, 2833
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O88410, P49682
CXCR3
CXCR3 Fusion Protein Ag25056
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions