missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14840-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is specific to CXCR7.
This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas.Spécification
CXCR7 | |
Polyclonal | |
Unconjugated | |
ACKR3 | |
Chemokine orphan receptor 1, CMKOR1, CXC R7, CXCR 7, CXCR7, G protein coupled receptor 159, GPR159, RDC 1, RDC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12778, 57007, 84348 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O89039, P25106, P56485 | |
ACKR3 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |