missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27242-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.Spécification
Cyclin A2 | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P20248 | |
CCN1, CCNA, CCNA2, Cyclin A, Cyclin A2 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
890 |
Western Blot | |
Unconjugated | |
CCNA2 | |
Liquid | |
CCNA2 | |
Cyclin A2 Fusion Protein Ag25666 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |