Learn More
ACROBiosystems Cynomolgus Glypican 3 / GPC3 Protein, His Tag
Marque:  ACROBiosystems GP3-C5225-1MG
Informations supplémentaires : Poids : 0.00001kg
 Afficher plus de versions de ce produit
Code produit. 15866867
  • 2421.11€ / 1mg
Date d’expédition estimée: 28-12-2021
Description
Description
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.
This protein carries a polyhistidine tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 24.6 kDa respectively. The N an
Spécification
Spécification
XP_005594665.1
HEK293
Cynomolgus
38.1 kDa and 24.6 kDa
1 mg
Gln 25 - His 559
GPC3,OCI5,Glypican-3,GTR2-2,MXR7,DGSX,SDYS ,SGB,SGBS,SGBS1
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>85% as determined by SDS-PAGE
Cynomolgus Glypican 3, His Tag (GP3-C5225) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # XP_005594665.1).
-20°C
Glypican 3
His Tag