missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus / Rhesus macaque B7-2 / CD86 Protein, Fc Tag

Marque:  ACROBiosystems CD6-C5254-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15876217

  • 1614.07€ / 1mg
Date d’expédition estimée: 29-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Cluster of Differentiation 86 (CD86) is also known as B-lymphocyte activation antigen B7-2, is a type I membrane protein that is a member of the immunoglobulin superfamily, and is constitutively expressed on interdigitating dendritic cells, Langerhans cells, peripheral blood dendritic cells, memory B cells, and germinal center B cells. Additionally, B72 is expressed at low levels on monocytes and can be upregulated through interferon γ. CD86 is the ligand for two different proteins on the T cell surface: CD28 (for autoregulation and intercellular association) and CTLA-4 (for attenuation of regulation and cellular disassociation). CD86 works in tandem with CD80 to prime T cells. Recent study has revealed that B7-2 promotes the generation of a mature APC repertoire and promotes APC function and survival. Furthermore, the B7 proteins are also involved in innate immune responses by activating NF-κB-signaling pathway in macrophages. CD86 thus is regarded as a promising candidate for immune therapy. CD86+ macrophages in Hodgkin lymphoma patients are an independent marker for potential nonresponse to firstline-therapy.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52 kDa. The protein migrates as 80-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>95% as determined by SDS-PAGE
Cynomolgus / Rhesus macaque B7-2, Fc Tag (CD6-C5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession # G7NXR4). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7-2 are homologus.
-20°C
B7-2
Fc Tag
HEK293
Cynomolgus / Rhesus macaque
52 kDa
1 mg
Ala 19 - His 240
CD86,B7-2,B70,CD28LG2,LAB72,MGC34413
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Vidéos
FDS
Documents
Certificats
Promotions

Promotions