Learn More
ACROBiosystems Cynomolgus / Rhesus macaque CTLA-4 Protein, Fc Tag
Marque: ACROBiosystems CT4-C5256-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
CTLA-4 (Cytotoxic T-Lymphocyte Antigen 4) is also known as CD152 (Cluster of differentiation 152), is a protein receptor that downregulates the immune system. CTLA4 is a member of the immunoglobulin superfamily, which is expressed on the surface of Helper T cells and transmits an inhibitory signal to T cells. The protein contains an extracellular V domain, a transmembrane domain, and a cytoplasmic tail. Alternate splice variants, encoding different isoforms. CTLA4 is similar to the T-cell co-stimulatory protein, CD28, and both molecules bind to CD80 and CD86, also called B7-1 and B7-2 respectively, on antigen-presenting cells. CTLA4 transmits an inhibitory signal to T cells, whereas CD28 transmits a stimulatory signal. Intracellular CTLA4 is also found in regulatory T cells and may be important to their function. Fusion proteins of CTLA4 and antibodies (CTLA4-Ig) have been used in clinical trials for rheumatoid arthritis.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.9 kDa. As a result of glycosylation, the protein migrates as 45-52 kDa under reducing (R) condition, and 90-105 kDa under non-reducing (NR) condition (SDS-PASpécification
XP_005574071.1 | |
1 mg | |
Ala 37 - Ser 160 | |
CTLA4,CD152 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
39.9 kDa | |
Cynomolgus / Rhesus macaque CTLA-4, Fc Tag (CT4-C5256) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession # G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus macaque CTLA-4 are homologus. | |
-20°C | |
CTLA-4 | |
Cynomolgus Monkey, Rhesus Macaque | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |