missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22208-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).Spécification
Cytokeratin 7 | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08729 | |
CK 7, CK7, Cytokeratin 7, K2C7, K7, keratin 7, KRT7, Sarcolectin, SCL, Type II keratin Kb7 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
3855 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
KRT7 | |
Liquid | |
KRT7 | |
Cytokeratin 7 Fusion Protein Ag17528 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |