missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYTSB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13851-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene belongs to the cytospin-A family. It is localized in the nucleus, and highly expressed in testis and some cancer cell lines. A chromosomal translocation involving this gene and platelet-derived growth factor receptor, beta gene (PDGFRB) may be a cause of juvenile myelomonocytic leukemia. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.Spécification
CYTSB | |
Polyclonal | |
Unconjugated | |
SPECC1 | |
cytospin B, CYTSB, HCMOGT 1, NSP, NSP5, Nuclear structure protein 5, SPECC1, Sperm antigen HCMOGT 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
303208, 432572, 92521 | |
-20°C | |
Liquid |
Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5M775, Q5SXY1 | |
Specc1 | |
CYTSB Fusion Protein Ag4822 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |