missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D2HGDH Mouse anti-Human, Mouse, Clone: 1E9B3, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66364-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes D-2hydroxyglutarate dehydrogenase, a mitochondrial enzyme belonging to the FAD-binding oxidoreductase/transferase type 4 family. This enzyme, which is most active in liver and kidney but also active in heart and brain, converts D-2-hydroxyglutarate to 2-ketoglutarate. Mutations in this gene are present in D-2-hydroxyglutaric aciduria, a rare recessive neurometabolic disorder causing developmental delay, epilepsy, hypotonia, and dysmorphic features.Spécification
D2HGDH | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CIM3, Q8N465 | |
D2HGDH | |
D2HGDH Fusion Protein Ag5036 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1E9B3 | |
Unconjugated | |
D2HGDH | |
D2HGD, D2HGDH | |
Mouse | |
Protein G | |
RUO | |
728294, 98314 | |
-20°C | |
Liquid |