missing translation for 'onlineSavingsMsg'
Learn More

DACH1 Mouse anti-Human, Mouse, Rat, Clone: 3B6D2, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 60082-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16827055

  • 325.38€ / 150µL
Expédition estimée: 29-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is a mouse monoclonal antibody raised against residues near the C terminus of human DACH1.

This gene encodes a member of the diaphanous subfamily of the formin family. Members of this family are involved in actin remodeling and regulate cell movement and adhesion. Mutations in this gene are associated with autosomal domit auditory neuropathy 1. Multiple transcript variants encoding different isoforms have been found for this gene.
Spécification

Spécification

DACH1
Monoclonal
0.83 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9QYB2, Q9UI36
DACH1
DACH1 Fusion Protein Ag4474
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
3B6D2
Unconjugated
DACH1
DACH, DACH1, Dachshund homolog 1
Mouse
Protein A
RUO
13134, 1602, 306096
-20°C
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions