missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX3Y Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | DDX3Y |
---|---|
Concentration | 0.43 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16812514
|
Proteintech
14041-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16802514
|
Proteintech
14041-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Polyclonal anti-DDX3Y antibodies (14041-1-AP) are produced by immunizing animals with C-terminus of DDX3Y and detects a 73 kDa band. As DDX3Y has a structural homolog with DDX3X, the antibody can recognize both.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases which are implicated in a number of cellular processes involving RNA binding and alteration of RNA secondary structure. DDX3Y is one member of DEAD box RNA helicases, encode by the gene locate on the chromosome Y in the azoospermia factor A region.Spécification
DDX3Y | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O15523, Q62095 | |
26900, 364073, 8653 | |
DDX3Y Fusion Protein Ag5110 | |
Primary | |
-20°C | |
DDX3Y |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DBY, DDX3Y | |
Ddx3y | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |