Learn More
DDX3Y Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14041-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Polyclonal anti-DDX3Y antibodies (14041-1-AP) are produced by immunizing animals with C-terminus of DDX3Y and detects a 73 kDa band. As DDX3Y has a structural homolog with DDX3X, the antibody can recognize both.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases which are implicated in a number of cellular processes involving RNA binding and alteration of RNA secondary structure. DDX3Y is one member of DEAD box RNA helicases, encode by the gene locate on the chromosome Y in the azoospermia factor A region.Spécification
DDX3Y | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15523, Q62095 | |
DBY, DDX3Y | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
26900, 364073, 8653 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
DDX3Y | |
Liquid | |
Ddx3y | |
DDX3Y Fusion Protein Ag5110 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |