missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX56 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | DDX56 |
---|---|
Concentration | 0.41 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16811535
|
Proteintech
21989-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 03-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16801535
|
Proteintech
21989-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 03-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene shows ATPase activity in the presence of polynucleotides and associates with nucleoplasmic 65S preribosomal particles. This gene may be involved in ribosome synthesis, most likely during assembly of the large 60S ribosomal subunit.Spécification
DDX56 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9D0R4, Q9NY93 | |
52513, 54606 | |
DDX56 Fusion Protein Ag17272 | |
Primary | |
-20°C | |
DDX56 |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DDX21, DDX26, DDX56, DEAD box protein 21, DEAD box protein 56, NOH61 | |
Ddx56 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |