missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHFR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15194-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, and is required for the synthesis of purines, thymidylic acid and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia.Spécification
DHFR | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00374, P00375, Q920D2 | |
DHFR, DHFRP1, dihydrofolate reductase | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
13361, 1719, 24312 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
DHFR | |
Liquid | |
DHFR | |
DHFR Fusion Protein Ag7331 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |