missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13889-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the Hedgehog family. The hedgehog gene family encodes signaling molecules that play an important role in regulating morphogenesis. This protein is predicted to be made as a precursor that is autocatalytically cleaved; the N-terminal portion is soluble and contains the signaling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the organism. Defects in this protein have been associated with partial gonadal dysgenesis (PGD) accompanied by minifascicular polyneuropathy. This protein may be involved in both male gonadal differentiation and perineurial development.Spécification
DHH | |
Polyclonal | |
Unconjugated | |
DHH | |
Desert hedgehog protein, DHH, HHG 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13363, 50846, 84380 | |
-20°C | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43323, Q61488 | |
DHH | |
DHH Fusion Protein Ag4850 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |