missing translation for 'onlineSavingsMsg'
Learn More

DHODH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14877-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16803764

  • 112.71€ / 20µL

Expédition estimée
02-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene catalyzes the fourth enzymatic step, the ubiquinone-mediated oxidation of dihydroorotate to orotate, in de novo pyrimidine biosynthesis. This protein is a mitochondrial protein located on the outer surface of the inner mitochondrial membrane.
Spécification

Spécification

DHODH
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35435, Q02127, Q63707
DHOdehase, DHODH, dihydroorotate dehydrogenase, Dihydroorotate oxidase
Rabbit
IgG
20 μL
-20°C
Primary
1723, 56749, 65156
Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Unconjugated
DHODH
Liquid
DHODH
DHODH Fusion Protein Ag6649
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Vidéos
FDS
Documents