Learn More
Diaion™ WK10(H), résine échangeuse d’ions, de type poreux faiblement acide, 2,5 mEq/ml sur polyméthacrylate
Produits pharmaceutiques, désalalination, récupération de métaux, antibiotiques, purifications du saccharose
Marque: Thermo Scientific Alfa Aesar 046666.A1
Code nomenclature Nacres: NC.05
Informations supplémentaires : Poids : 1.00000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Billes | |
Weakly acidic porous type, 2.5meq/mL on polymethacrylate | |
1 kg |
Diaion™ WK10 (H), ion exchange resin | |
300 to 1180μm |