missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIO2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26513-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene belongs to the iodothyronine deiodinase family. It activates thyroid hormone by converting the prohormone thyroxine by outer ring deiodination to bioactive 3,3′,5-triiodothyronine residues encoded by the UGA codon, which normally signals translation termination. The 3′ UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence , which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Alternative splicing results in multiple transcript variants encoding different isoforms.Spécification
DIO2 | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q92813, Q9Z1Y9 | |
5DII, D2, DIO2, DIOII, ITDI2, SelY, TXDI2, Type 2 DI, Type II 5 deiodinase | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
13371, 1734 |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
DIO2 | |
Liquid | |
DIO2 | |
DIO2 Fusion Protein Ag24119 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |