missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIRAS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15557-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
DIRAS2 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.Spécification
DIRAS2 | |
Polyclonal | |
Unconjugated | |
DIRAS2 | |
Di Ras2, DIRAS2, GTP binding protein Di Ras2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291006, 54769, 68203 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5PR73, Q96HU8 | |
Diras2 | |
DIRAS2 Fusion Protein Ag7926 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |