missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIXDC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13816-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.Spécification
DIXDC1 | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q155Q3, Q2VUH7, Q80Y83 | |
CCD1, Coiled coil DIX1, Coiled coil protein DIX1, DIX domain containing 1, DIXDC1, Dixin, KIAA1735 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
330938, 363062, 85458 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
DIXDC1 | |
Liquid | |
Dixdc1 | |
DIXDC1 Fusion Protein Ag4904 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |