missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12338-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of a subclass of the HSP40/DnaJ protein family. Members of this family of proteins are associated with complex assembly, protein folding, and export. Two transcript variants encoding distinct isoforms have been identified for this gene.Spécification
DNAJC12 | |
Polyclonal | |
Unconjugated | |
DNAJC12 | |
DNAJC12, J domain containing protein 1, JDP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
30045, 56521, 619393 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q925T0, Q9R022, Q9UKB3 | |
Dnajc12 | |
DNAJC12 Fusion Protein Ag2989 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |