Learn More
STEMCELL Technologies DNase I
For digestion of DNA
Marque:  STEMCELL Technologies 07469
Informations supplémentaires : Poids : 0.00001kg
Code produit. 15964601
  • 105.07€ / 25mg
Date d’expédition estimée: 30-11-2021
Description
Description
  • Deoxyribonuclease I (DNase I) is an endonuclease consisting of a single glycosylated polypeptide chain with two disulfide bonds
  • DNase is often included in tissue dissociation protocols to digest DNA that has leaked into the dissociation medium as a result of cell damage
  • DNase I preferentially cleaves phosphodiester linkages adjacent to pyrimidine nucleotides in both single- and double-stranded DNA, yielding polynucleotides with 5'-phosphate and 3'-hydroxyl groups (Bernardi et al.) DNase I has been used for the digestion of human tissues such as microglia (Klegeris & McGeer), cartilage (Dunham & Koch), colon (Fukushima & Fiocchi), epithelium (Fukushima & Fiocchi), liver (Vatakis et al.), lung (Fujino et al.), neural (Fuja et al.), and stem cells (Kusuma et al.)
Spécification
Spécification
25 mg
Digestion of DNA